Transformation buffer-2, pH 7.5, Thermo Scientific
125ml
Supplier: Thermo Scientific Chemicals
Fournisseur #: J63293-EQE
Transformation buffer-2, pH 7.5, Thermo Scientific
Catalog # CAAAJ63293-EQE
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
For transformation of competent cells
Spécifications
Spécifications
- Taille:125ml



