RNA sample loading buffer, Thermo Scientific
2 mL
Supplier: Thermo Scientific Chemicals
Fournisseur #: J62832-S0
RNA sample loading buffer, Thermo Scientific
Catalog # CAAAJ62832-S0
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
MDL No.: MFCD00213041
Spécifications
Spécifications
- Taille:2 mL
- Cat. No.:CAAAJ62832-S0
- MDL No.:MFCD00213041



