GTE buffer
250 mL
Supplier: Thermo Scientific Chemicals
Supplier #: J62597-AK
Jump to:
Product Details & Documents
Storage temperature: 2 to 8°C
- Suitable for miniprep and DNA purification
Contains 25 mM Tris, 50 mM glucose, and 10 mM EDTA.
Specifications
Specifications
- Taille:250 mL



