HEPPSO, 0.2 M Buffer Solution, pH 8.0, Thermo Scientific
100 mL
HEPPSO, 0.2 M Buffer Solution, pH 8.0, Thermo Scientific
Catalog # CAAAJ62529-AE
CAS Number:
Jump to:
Discontinued
Product Details & Documents
Liquid
CAS No.: 68399-78-0
Physical Form: Liquid
MDL No.: MFCD00038351
Spécifications
Spécifications
- Taille:100 mL
- Cat. No.:CAAAJ62529-AE
- MDL No.:MFCD00038351



