Rapamycin ≥99%
200mg
  CAAAJ62473-EX3
 :  J62473-EX3
Rapamycin ≥99%
  CAAAJ62473-EX3
 :  53123-88-9
Rapamycin ≥99%
200mg
 :  53123-88-9
939,22 $
Each (200mg)
Blocks cytokine-mediated signal transduction pathways
Blocks cytokine-mediated signal transduction pathways
Blocks cytokine-mediated signal transduction pathways
 :  (3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone
  •  :53123-88-9
  •  :C₅₁H₇₉NO₁₃
  •  :MFCD00867594
  •  :914.19 g/mol
  •  :14,8114
  •  :Congélateur
  • Taille:
    200mg