ADA, 0.2 M Buffer Solutions, Thermo Scientific
ADA buffer solution, 0.2 M, 250 mL, Glass bottle, 7.5
Supplier: Thermo Scientific Chemicals
: J62424-AK
ADA, 0.2 M Buffer Solutions, Thermo Scientific
Catalog # CAAAJ62424-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
N-(2-Acetimido)iminodiacetic acid monosodium salt (ADA) buffer solutions PH values 6.0 to 7.5.
CAS No.: 7415-22-7
Appearance: Colorless
MDL No.: MFCD00065483
: Storage Temperature: 2 to 8°C
Spécifications
Spécifications
- Concentration:0.2 M
- Type de conditionnement:Glass bottle
- Désignation:ADA buffer solution
- Taille:250 mL
- pH:7.5
- Cat. No.:CAAAJ62424-AK
- MDL No.:MFCD00065483



