MOPS, 0.5 M Buffer Solutions, Thermo Scientific
MOPS buffer solution, 0.5 M, 100 mL, 8.5
MOPS, 0.5 M Buffer Solutions, Thermo Scientific
Catalog # CAAAJ62368-AE
CAS Number:
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
Liquid
CAS No.: 1132-61-2
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00006183
Specifications
Specifications
- Concentration:0.5 M
- Désignation:MOPS buffer solution
- Taille:100 mL
- pH:8.5
- Cat. No.:CAAAJ62368-AE
- Odour/scent:Odorless
- Storage temperature:2°C to 8°C



