MOBS, 0.2 M Buffer Solution, pH 8.5, Thermo Scientific
250 mL
MOBS, 0.2 M Buffer Solution, pH 8.5, Thermo Scientific
Catalog # CAAAJ62203-AK
CAS Number:
Jump to:
Discontinued
Product Details & Documents
Liquid
CAS No.: 115724-21-5
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00674916
Spécifications
Spécifications
- Taille:250 mL
- Cat. No.:CAAAJ62203-AK
- MDL No.:MFCD00674916



