LDS-sample buffer (4X), non-reducing, Thermo Scientific
50 mL
Supplier: Thermo Scientific Chemicals
Fournisseur #: J61894-AD
LDS-sample buffer (4X), non-reducing, Thermo Scientific
Catalog # CAAAJ61894-AD
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
LDS-sample buffer (4X), non-reducing is used in SDS-polyacrylamide gel electrophoresis (SDS-PAGE). It is used in denaturing gels and is compatible with both coomassie and silver staining and Western blotting applications.
Physical Form: Liquid
Appearance: Blue
This buffer contains: 988 mM Tris-HCl, 2.04 mM EDTA, 8% LDS, 40% glycerol, 0.88% Coomassie blue and 0.7 mM phenol red at pH 8.5.
Spécifications
Spécifications
- Taille:50 mL
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



