Succinate, 0.2 M Buffer Solution, pH 6.0, Thermo Scientific
250 mL
Supplier: Thermo Scientific Chemicals
Supplier #: J61853-AK
Succinate, 0.2 M Buffer Solution, pH 6.0, Thermo Scientific
Catalog # CAAAJ61853-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
CAS No.: 150-90-3
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00002789
Spécifications
Spécifications
- Taille:250 mL
- Cat. No.:CAAAJ61853-AK
- MDL No.:MFCD00002789
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



