EPPS, 0.2 M Buffer Solutions, Thermo Scientific
EPPS buffer solution, 0.2 M, 250 mL, Glass bottle, 8.5
Supplier: Thermo Scientific Chemicals
Supplier #: J61476-AK
EPPS, 0.2 M Buffer Solutions, Thermo Scientific
Catalog # CAAAJ61476-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
CAS No.: 16052-06-5
Physical Form: Liquid
MDL No.: MFCD00006160
Specifications
Specifications
- Concentration:0.2 M
- Type de conditionnement:Glass bottle
- Désignation:EPPS buffer solution
- Taille:250 mL
- pH:8.5
- Cat. No.:CAAAJ61476-AK
- MDL No.:MFCD00006160



