EPPS, 0.2 M Buffer Solutions, Thermo Scientific
EPPS buffer solution, 0.2 M, 250 mL, Glass bottle, 8.0
Supplier: Thermo Scientific Chemicals
Fournisseur #: J61296-AK
EPPS, 0.2 M Buffer Solutions, Thermo Scientific
Catalog # CAAAJ61296-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
CAS No.: 16052-06-5
Physical Form: Liquid
MDL No.: MFCD00006160
Spécifications
Spécifications
- Concentration:0.2 M
- Type de conditionnement:Glass bottle
- Désignation:EPPS buffer solution
- Taille:250 mL
- pH:8.0
- Cat. No.:CAAAJ61296-AK
- MDL No.:MFCD00006160
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



