ADA, 0.2 M Buffer Solutions, Thermo Scientific
ADA buffer solution, 0.2 M, 250 mL, Glass bottle, 6.5
: J61259-AK
N-(2-Acetimido)iminodiacetic acid monosodium salt (ADA) buffer solutions PH values 6.0 to 7.5.
CAS No.: 7415-22-7
Appearance: Colorless
MDL No.: MFCD00065483
: Storage Temperature: 2 to 8°C
- Concentration:0.2 M
- Type de conditionnement:Glass bottle
- Désignation:ADA buffer solution
- Taille:250 mL
- pH:6.5
- Cat. No.:CAAAJ61259-AK
- MDL No.:MFCD00065483



