MOPS, 0.5 M Buffer Solutions, Thermo Scientific
MOPS buffer solution, 0.5 M, 250 mL, 8.0
Supplier: Thermo Scientific Chemicals
: J61236-AK
MOPS, 0.5 M Buffer Solutions, Thermo Scientific
Catalog # CAAAJ61236-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
CAS No.: 1132-61-2
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00006183
Spécifications
Spécifications
- Concentration:0.5 M
- Désignation:MOPS buffer solution
- Taille:250 mL
- pH:8.0
- Cat. No.:CAAAJ61236-AK
- Odour/scent:Odorless
- Storage temperature:2°C to 8°C
- MDL No.:MFCD00006183



