Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Tricine-SDS Sample Buffer (2X), reducing, Thermo Scientific

Tricine-SDS Sample Buffer (2X), reducing, Thermo Scientific

25 mL
  CAAAJ61042-AC
Supplier #:  J61042-AC
Tricine-SDS Sample Buffer (2X), reducing, Thermo Scientific
  CAAAJ61042-AC
 :  
Tricine-SDS Sample Buffer (2X), reducing, Thermo Scientific

25 mL

 :  
 
$125.66
   
Each (25ml)
Minimum order quantity is 1
 

Liquid

Physical Form: Liquid
Appearance: Blue

 

 

  • Taille:
    25 mL
  • Cat. No.:
    CAAAJ61042-AC

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.