MOPS, 0.5 M Buffer Solutions, Thermo Scientific
MOPS buffer solution, 0.5 M, 250 mL, 9.0
MOPS, 0.5 M Buffer Solutions, Thermo Scientific
Catalog # CAAAJ60922-AK
CAS Number:
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
3-MORPHOLINOPROPANESULFONIC AC Catalog #TCM0707-250G
Product TCM0707-250G is a replacement for CAAAJ60922-AK and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Liquid
CAS No.: 1132-61-2
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00006183
Specifications
Specifications
- Concentration:0.5 M
- Désignation:MOPS buffer solution
- Taille:250 mL
- pH:9.0
- Cat. No.:CAAAJ60922-AK
- Odour/scent:Odorless
- Storage temperature:2°C to 8°C
- MDL No.:MFCD00006183
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



