AMPD, 0.2 M Buffer Solutions, Thermo Scientific
AMPD buffer solution, 0.2 M, 250 mL, 9.5
Supplier: Thermo Scientific Chemicals
Supplier #: J60916-AK
AMPD, 0.2 M Buffer Solutions, Thermo Scientific
Catalog # CAAAJ60916-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
2-Amino-2-methyl-1,3-propanediol buffer, colorless and odorless.
CAS No.: 115-69-5
MDL No.: MFCD00004678
: Storage Temperature: 2 to 8 °C
Specifications
Specifications
- Concentration:0.2 M
- Désignation:AMPD buffer solution
- Taille:250 mL
- pH:9.5
- Cat. No.:CAAAJ60916-AK
- MDL No.:MFCD00004678



