Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

SSC buffer (20X), Thermo Scientific

4 L
  CAAAJ60839-K7
 :  J60839-K7
SSC buffer (20X), Thermo Scientific
  CAAAJ60839-K7
 :  
SSC buffer (20X), Thermo Scientific

4 L

 :  
 
276,48 $
   
Each (4l)
 
 

Liquid

Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00132371

 

 

 

  • Taille:
    4 L
  • Cat. No.:
    CAAAJ60839-K7
  • MDL No.:
    MFCD00132371

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.