Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
CHAPS Lysis Buffer (2X), Thermo Scientific

CHAPS Lysis Buffer (2X), Thermo Scientific

25 mL
Catalog # CAAAJ60812-AC
Supplier #:  J60812-AC
CHAPS Lysis Buffer (2X), Thermo Scientific
Catalog # CAAAJ60812-AC
CAS Number:  
CHAPS Lysis Buffer (2X), Thermo Scientific

25 mL

CAS Number:  
Price
$207.11
Log in for availability
Each (25ml)
Minimum order quantity is 1
Product Details & Documents

Liquid

Physical Form: Liquid
Appearance: Clear liquid
MDL No.: MFCD00282866

Specifications

Specifications

  • Taille:
    25 mL
  • Cat. No.:
    CAAAJ60812-AC
  • MDL No.:
    MFCD00282866

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.