TNT Buffer Solution, Thermo Scientific
1 L
Supplier: Thermo Scientific Chemicals
Supplier #: J60622-K2
TNT Buffer Solution, Thermo Scientific
Catalog # CAAAJ60622-K2
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
Physical Form: Liquid
Spécifications
Spécifications
- Taille:1 L
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



