CHAPS Lysis Buffer, Thermo Scientific
50 mL
Supplier: Thermo Scientific Chemicals
Fournisseur #: J60580-AD
CHAPS Lysis Buffer, Thermo Scientific
Catalog # CAAAJ60580-AD
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
Physical Form: Liquid
Appearance: Clear liquid
MDL No.: MFCD00282866
Applications: For cell lysis and protein extraction
Spécifications
Spécifications
- Taille:50 mL
- Cat. No.:CAAAJ60580-AD
- MDL No.:MFCD00282866



