EPPS, 0.2 M Buffer Solutions, Thermo Scientific
EPPS buffer solution, 0.2 M, 100 mL, Glass bottle, 7.5
Supplier: Thermo Scientific Chemicals
: J60511-AE
EPPS, 0.2 M Buffer Solutions, Thermo Scientific
Catalog # CAAAJ60511-AE
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
CAS No.: 16052-06-5
Physical Form: Liquid
MDL No.: MFCD00006160
Spécifications
Spécifications
- Concentration:0.2 M
- Type de conditionnement:Glass bottle
- Désignation:EPPS buffer solution
- Taille:100 mL
- pH:7.5
- Cat. No.:CAAAJ60511-AE
- MDL No.:MFCD00006160



