3,3',5,5'-Tétraméthylbenzidine solution, sensibilité standard prêt-à-l'emploi, précipitant
500 mL
Supplier: Thermo Scientific Chemicals
Supplier #: J60461-AP
3,3',5,5'-Tétraméthylbenzidine solution, sensibilité standard prêt-à-l'emploi, précipitant
Catalog # CAAAJ60461-AP
Supplier: Thermo Scientific Chemicals
CAS Number: 54827-17-7
Jump to:
Product Details & Documents
Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result
Liquid
Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result
Synonymes: TMB
- CAS Number:54827-17-7
- Formule:C₁₆H₂₀N₂
- Numéro MDL:MFCD00007748
- Molecular Weight:240.35 g/mol
- Température de conservation:Température ambiante
- L'EINECS:259-364-6
- ONU:2810
- MARC:6.1,III
- MARC:"6.1",III
Spécifications
Spécifications
- Taille:500 mL
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



