Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
FSB buffer, Thermo Scientific

FSB buffer, Thermo Scientific

250 mL
Catalog # CAAAJ60262-AK
Supplier #:  J60262-AK
FSB buffer, Thermo Scientific
Catalog # CAAAJ60262-AK
CAS Number:  
FSB buffer, Thermo Scientific

250 mL

CAS Number:  
Price
$480.04
Log in pour la disponibilité
Each (250ml)
Minimum order quantity is 1
Product Details & Documents

Liquid

Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
For frozen storage of competent cells

Spécifications

Spécifications

  • Taille:
    250 mL

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.