Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
TBE urea sample buffer (2X), Thermo Scientific | Avantor

TBE urea sample buffer (2X), Thermo Scientific

10 mL
Catalog # CAAAJ60186-AB
Fournisseur #:  J60186-AB
TBE urea sample buffer (2X), Thermo Scientific
Catalog # CAAAJ60186-AB
CAS Number:  
TBE urea sample buffer (2X), Thermo Scientific

10 mL

CAS Number:  
Price
$150.79
Log in pour la disponibilité
Each (10ml)
La quantité minimale de commande est de 1
Product Details & Documents

Liquid

Physical Form: Liquid
Appearance: Blue

Spécifications

Spécifications

  • Taille:
    10 mL

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.