Denaturing lysis buffer, Thermo Scientific
250 mL
Supplier: Thermo Scientific Chemicals
Fournisseur #: J60132-AK
Denaturing lysis buffer, Thermo Scientific
Catalog # CAAAJ60132-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
Physical Form: Liquid
Appearance: Clear liquid
pH: 7.4
Spécifications
Spécifications
- Taille:250 mL
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



