Solid Caps, GL-Threads, Polypropylene, Chemglass
Solid Caps, Yellow, GL45
Catalog # 30623-024
Supplier: Chemglass
Supplier #: CLS-1480-18
Jump to:
Product Details & Documents
These solid polypropylene caps can be autoclaved in steam to 140 °C.
Specifications
- Color:Yellow
- Désignation:Solid Caps
- Epaisseur:GL45