Solid Caps, GL-Threads, Polypropylene, Chemglass
Solid Caps, Green, GL45
Catalog # 30623-018
Supplier: Chemglass
Fournisseur #: CLS-1480-15
Jump to:
Product Details & Documents
These solid polypropylene caps can be autoclaved in steam to 140 °C.
Spécifications
- Color:Green
- Désignation:Solid Caps
- Epaisseur:GL45