Solid Caps, GL-Threads, Polypropylene, Chemglass
Solid Caps, Gray, GL45
Catalog # 30623-016
Supplier: Chemglass
Fournisseur #: CLS-1480-14
Jump to:
Product Details & Documents
These solid polypropylene caps can be autoclaved in steam to 140 °C.
Spécifications
- Color:Gray
- Désignation:Solid Caps
- Epaisseur:GL45