Solid Caps, GL-Threads, Polypropylene, Chemglass
Solid Caps, Schott Blue, GL45
30623-014
: CLS-1480-13SC
These solid polypropylene caps can be autoclaved in steam to 140 °C.
- Color:Schott Blue
- Désignation:Solid Caps
- Epaisseur:GL45
These solid polypropylene caps can be autoclaved in steam to 140 °C.