Solid Caps, GL-Threads, Polypropylene, Chemglass
Solid Caps, Blue, GL32
Catalog # 30623-010
Supplier: Chemglass
Supplier #: CLS-1480-11
Jump to:
Product Details & Documents
These solid polypropylene caps can be autoclaved in steam to 140 °C.
Spécifications
- Color:Blue
- Désignation:Solid Caps
- Epaisseur:GL32