Solid Caps, GL-Threads, Polypropylene, Chemglass
Solid Caps, Blue, GL25
30623-008
: CLS-1480-10
These solid polypropylene caps can be autoclaved in steam to 140 °C.
- Color:Blue
- Désignation:Solid Caps
- Epaisseur:GL25
These solid polypropylene caps can be autoclaved in steam to 140 °C.