VWR® Buffer Solutions
VWR® Buffer Solutions
Catalog # CA11021-938
VWR® Buffer Solutions
Catalog # CA11021-938
Supplier:  
CAS Number:  
VWR® Buffer Solutions
VWR® Buffer Solutions
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BUFFER SOL PH 6.00 +0.01 2 4L Catalog #CAAA42418-K7
Product CAAA42418-K7 is similar to CA11021-938 but manufacturer and specifications may vary.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These buffer solutions are for calibrating pH meters
They are traceable to NIST standards.