Stir Bar, Triangular, PTFE, Dynalon | Avantor
Stir Bar, Triangular, PTFE, Dynalon
35 mm (1⅜"), 9 mm (1/3"), PTFE, 0.0225 lbs.
Catalog # 30620-486
Supplier:   Dynalon
Fournisseur #:  302826
Stir Bar, Triangular, PTFE, Dynalon
Catalog # 30620-486
Supplier:  Dynalon
Stir Bar, Triangular, PTFE, Dynalon
Stir Bar, Triangular, PTFE, Dynalon
35 mm (1⅜"), 9 mm (1/3"), PTFE, 0.0225 lbs.
Price
$275.14
Log in pour la disponibilité
Case of 12
Product Details & Documents
Triangular shape provides the best possible spinning action and most efficient mixing of all stir bars
Powerful permanent Alnico V magnetic cores. PTFE offers excellent chemical resistance. Always store in pairs to maintain magnetic strength.
Spécifications
  • Revêtement:
    PTFE
  • Ø:
    9 mm (1/3")
  • Longueur:
    35 mm (1⅜")
  • Weight:
    0.0225 lbs.