Sample Cups, Polystyrene, Dynalon
4.0 ml, 39 mm
Catalog # 30620-272
Supplier: Dynalon
Fournisseur #: 202134
Jump to:
Product Details & Documents
Clear polystyrene cups for use with older auto analyzers.
Also useful for general micro use. Conical bottom. 26mm height. LDPE Stopper available
Spécifications
- Capacité:4.0 ml
- Hauteur:39 mm