Sample Cups, Polystyrene, Dynalon
1.5 ml, 24 mm
Catalog # 30620-268
Supplier: Dynalon
Fournisseur #: 202114
Jump to:
Product Details & Documents
Clear polystyrene cups for use with older auto analyzers.
Also useful for general micro use. Conical bottom. 26mm height. LDPE Stopper available
Spécifications
- Capacité:1.5 ml
- Hauteur:24 mm