Accessories for Eppendorf® Microcentrifuges Model 5430, Eppendorf
Adapter, for 2 conical tubes, skirted 50 ml, for rotor S-4-72, 2 pieces
Catalog # CA11017-448
Supplier: Eppendorf
Fournisseur #: 5804785002
Accessories for Eppendorf® Microcentrifuges Model 5430, Eppendorf
Catalog # CA11017-448
Supplier: Eppendorf
Jump to:
Product Details & Documents
Spécifications
- Désignation:Adapter, for 2 conical tubes, skirted 50 ml, for rotor S-4-72, 2 pieces
- Cat. No.:CA11017-448
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Rotor key, 5 mmRotor key, 5 mmCA53513-248Centrifuges 5417 C/R, 5418/R, 5424/R, 5430/R and 5702/R/RHLog in pour la disponibilitéEach - $42.30