Accessories for Eppendorf® Microcentrifuges Model 5430, Eppendorf
Adapter, for 26 tubes 1.5/2.0 ml, for rotor S-4-72, 2 pieces
Catalog # CA11017-434
   Low Manufacturing Impact
Supplier:   Eppendorf
 :  5804794001
Accessories for Eppendorf® Microcentrifuges Model 5430, Eppendorf
Catalog # CA11017-434
Supplier:  Eppendorf
   Low Manufacturing Impact
Accessories for Eppendorf® Microcentrifuges Model 5430, Eppendorf
Accessories for Eppendorf® Microcentrifuges Model 5430, Eppendorf
Adapter, for 26 tubes 1.5/2.0 ml, for rotor S-4-72, 2 pieces
Price
$617.00
Log in pour la disponibilité
Pack of 2
Product Details & Documents
Spécifications
  • Désignation:
    Adapter, for 26 tubes 1.5/2.0 ml, for rotor S-4-72, 2 pieces
  • Cat. No.:
    CA11017-434
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Rotor key, 5 mm
    Rotor key, 5 mm
    CA53513-248
    Centrifuges 5417 C/R, 5418/R, 5424/R, 5430/R and 5702/R/RH
    Log in pour la disponibilité
    Each - $42.30