Accessories for Eppendorf® Microcentrifuges Model 5430, Eppendorf
Adapter, for 20 round bottom tubes 5.5 - 12 ml, 2 pieces, for rotor S-4-104
Catalog # CA11017-406
Supplier: Eppendorf
Fournisseur #: 5825736001
Accessories for Eppendorf® Microcentrifuges Model 5430, Eppendorf
Catalog # CA11017-406
Supplier: Eppendorf
Jump to:
Product Details & Documents
Spécifications
- Désignation:Adapter, for 20 round bottom tubes 5.5 - 12 ml, 2 pieces, for rotor S-4-104
- Cat. No.:CA11017-406
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Informations sur le produit
- Catalog #For Use WithAvailabilityPrice
- Rotor key, 5 mmRotor key, 5 mmCA53513-248Centrifuges 5417 C/R, 5418/R, 5424/R, 5430/R and 5702/R/RHLog in pour la disponibilitéEach - $42.30