Hydroxylammonium chlorure solution 100 g/l
100 ml
Supplier: Hach
Supplier #: 81842
Hydroxylammonium chlorure solution 100 g/l
Catalog # CA11016-776
Supplier: Hach
CAS Number: 5470-11-1
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
For determination of volatile acids by the esterification method
Hach method 8196 (100 tests).
Synonyms: Hydroxylamine chlorhydrate
- CAS Number:5470-11-1
- Formula:NH₂OH·HCl
- MDL Number:MFCD00051089
- Molecular Weight:69.49 g/mol
- Storage Temperature:Température ambiante
Specifications
Specifications
- Taille:100 ml



