Test Kits, COD, TNT plus®
Test Kits, COD, TNT plus®
COD TNTplus™, ULR (digestion required), 25 Tubes/pack, Reactor digestion, 1 - 60 mg/L COD
  CA11014-646
 :   Hach
Supplier #:  TNT820
Test Kits, COD, TNT plus®
  CA11014-646
 :  Hach
 :  
Test Kits, COD, TNT plus®
Test Kits, COD, TNT plus®
COD TNTplus™, ULR (digestion required), 25 Tubes/pack, Reactor digestion, 1 - 60 mg/L COD
 :  
$190.67
Pack of 25
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are color coded by parameter and range
  • No reagent blank necessary
TNTplus™ works only with DR 6000™ UV-VIS, DR 3900™ Benchtop, and DR 1900™ Portable spectrophotometers.
  • Méthode chimique:
    Reactor digestion
  • Désignation:
    COD TNTplus™, ULR (digestion required)
  • Taille:
    25 Tubes/pack
  • Range:
    1 - 60 mg/L COD