SINGLET™ Single-Use pH Buffer, Combination Pack, Hach
±0.02, 10.01
Supplier: Hach
Supplier #: 2770220
SINGLET™ Single-Use pH Buffer, Combination Pack, Hach
Catalog # CA11014-256
Supplier: Hach
CAS Number:
Jump to:
Product Details & Documents
Single-use standard solutions eliminate the need to prepare standardized buffer solutions for pH meter calibration
- Ideal for on-site analysis
- Easy to use
- Fresh and reliable
You can have fresh pH buffer solutions without having to purchase and carry deionized or demineralized water. The convenient foil-pouch packaging maintains freshness, and keeps the solutions contaminant-free and ready to travel with you anywhere.
Ordering information: Single-Use pH Buffer Combination Packs are packaged with 20 pouches with 10 of each pH listed per pack.
Spécifications
Spécifications
- pH:10.01
- pH Accuracy:±0.02
- Cat. No.:CA11014-256
- Concentration:10.01 pH
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



