SINGLET™ Single-Use pH Buffer, Combination Pack, Hach
Combination Pack, ±0.02, 4.01/7.00
  CA11014-254
 :   Hach
 :  2769920
SINGLET™ Single-Use pH Buffer, Combination Pack, Hach
  CA11014-254
 :  Hach
 :  
SINGLET™ Single-Use pH Buffer, Combination Pack, Hach
SINGLET™ Single-Use pH Buffer, Combination Pack, Hach
Combination Pack, ±0.02, 4.01/7.00
 :  
$138.75
Pack of 10
Single-use standard solutions eliminate the need to prepare standardized buffer solutions for pH meter calibration
  • Ideal for on-site analysis
  • Easy to use
  • Fresh and reliable
You can have fresh pH buffer solutions without having to purchase and carry deionized or demineralized water. The convenient foil-pouch packaging maintains freshness, and keeps the solutions contaminant-free and ready to travel with you anywhere.
 : Single-Use pH Buffer Combination Packs are packaged with 20 pouches with 10 of each pH listed per pack.
  • Désignation:
    Combination Pack
  • pH:
    4.01/7.00
  • pH Accuracy:
    ±0.02