SINGLET™ Single-Use pH Buffer, Combination Pack, Hach
Combination Pack, ±0.02, 4.01/7.00
CA11014-254
: Hach
: 2769920
Single-use standard solutions eliminate the need to prepare standardized buffer solutions for pH meter calibration
- Ideal for on-site analysis
- Easy to use
- Fresh and reliable
You can have fresh pH buffer solutions without having to purchase and carry deionized or demineralized water. The convenient foil-pouch packaging maintains freshness, and keeps the solutions contaminant-free and ready to travel with you anywhere.
: Single-Use pH Buffer Combination Packs are packaged with 20 pouches with 10 of each pH listed per pack.
- Désignation:Combination Pack
- pH:4.01/7.00
- pH Accuracy:±0.02