SINGLET™ Single-Use pH Buffer, Combination Pack, Hach
±0.02, 4.01
Supplier: Hach
: 2770020
SINGLET™ Single-Use pH Buffer, Combination Pack, Hach
Catalog # CA11014-252
Supplier: Hach
CAS Number:
Jump to:
Product Details & Documents
Single-use standard solutions eliminate the need to prepare standardized buffer solutions for pH meter calibration
- Ideal for on-site analysis
- Easy to use
- Fresh and reliable
You can have fresh pH buffer solutions without having to purchase and carry deionized or demineralized water. The convenient foil-pouch packaging maintains freshness, and keeps the solutions contaminant-free and ready to travel with you anywhere.
: Single-Use pH Buffer Combination Packs are packaged with 20 pouches with 10 of each pH listed per pack.
Spécifications
Spécifications
- pH:4.01
- pH Accuracy:±0.02
- Cat. No.:CA11014-252
- Concentration:4.01 pH



