Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Corning® Polypropylene Funnel, Corning

Short Stem Funnel, 75 mm, PP, 110 mm
Catalog # 30617-112
Supplier:   Corning
 :  6120P-75
Corning® Polypropylene Funnel, Corning
Catalog # 30617-112
Supplier:  Corning
Corning® Polypropylene Funnel, Corning
Corning® Polypropylene Funnel, Corning

Short Stem Funnel, 75 mm, PP, 110 mm

Price
130,90 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 12 Each
Product Details & Documents

Funnels, made of durable PP, translucent.

  • Very good chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper

Allow safe and easy transfers of solids or liquids from one vessel to another.

Spécifications
  • Ø:
    75 mm
  • Désignation:
    Short Stem Funnel
  • Longueur:
    110 mm
  • Matière:
    PP