Corning® Polypropylene Funnel, Corning
Short Stem Funnel, 50 mm, PP, 85 mm
  30617-110
 :   Corning
 :  6120P-50
Corning® Polypropylene Funnel, Corning
  30617-110
 :  Corning
Corning® Polypropylene Funnel, Corning
Corning® Polypropylene Funnel, Corning
Short Stem Funnel, 50 mm, PP, 85 mm
$218.00
Case of 24 Each
Funnels, made of durable PP, translucent.
  • Very good chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper
Allow safe and easy transfers of solids or liquids from one vessel to another.
  • Ø:
    50 mm
  • Désignation:
    Short Stem Funnel
  • Longueur:
    85 mm
  • Matière:
    PP