Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Corning® Polypropylene Funnel, Corning

Corning® Polypropylene Funnel, Corning

Short Stem Funnel, 30 mm, PP, 45 mm
Catalog # 30617-106
Supplier:   Corning
Supplier #:  6120P-30
Corning® Polypropylene Funnel, Corning
Catalog # 30617-106
Supplier:  Corning
Corning® Polypropylene Funnel, Corning
Corning® Polypropylene Funnel, Corning

Short Stem Funnel, 30 mm, PP, 45 mm

Price
$131.75
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 24 Each
Product Details & Documents

Funnels, made of durable PP, translucent.

  • Very good chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper

Allow safe and easy transfers of solids or liquids from one vessel to another.

Spécifications
  • Ø:
    30 mm
  • Désignation:
    Short Stem Funnel
  • Longueur:
    45 mm
  • Matière:
    PP
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.