Corning® Polypropylene Erlenmeyer Flasks, Corning
1000 ml, 200 ml, GL-52
Supplier: Corning
Supplier #: 4985P-1L
Jump to:
Product Details & Documents
PP, translucent.
- Very good chemical resistance
- Very good temperature resistance
With blue, easy to read graduations. Autoclavable. The flasks feature a stopper bed and can also be sealed with standard ground joint polypropylene stoppers.
Specifications
- Intervalle de graduation:200 ml
- Capacité:1000 ml
- Epaisseur:GL-52