Corning® Polypropylene Erlenmeyer Flasks, Corning
50 ml, 10 ml, GL-40
Supplier #: 4985P-50
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
FLASK ERLENMEYER 50 ML PP PK6 Catalog #83008-938
Product 83008-938 is similar to 30617-086 but manufacturer and specifications may vary.
Product Details & Documents
PP, translucent.
- Very good chemical resistance
- Very good temperature resistance
With blue, easy to read graduations. Autoclavable. The flasks feature a stopper bed and can also be sealed with standard ground joint polypropylene stoppers.
Specifications
- Intervalle de graduation:10 ml
- Capacité:50 ml
- Epaisseur:GL-40