Corning® Polypropylene Bottles, Corning
Wide Mouth, 500 ml, PP, 45
Supplier: Corning
Supplier #: 1300P-500W
Jump to:
Product Details & Documents
Bottles are manufactured from low density polyethylene and are offered with a narrow or wide mouth design.
- Hollow glass stopper style
- Reusable
- Non sterile
The cap and dispensing tube are manufactured from polypropylene to optimize backflow of the solution. The design permits minimal turbulence of the liquid in the bottle.
Specifications
- Capacité:500 ml
- Désignation:Wide Mouth
- Matière:PP
- GL Thread Size:45