Corning® Polypropylene Bottles, Corning
Corning® Polypropylene Bottles, Corning
Wide Mouth, 500 ml, PP, 45
Catalog # 30617-072
Supplier:   Corning
Supplier #:  1300P-500W
Corning® Polypropylene Bottles, Corning
Catalog # 30617-072
Supplier:  Corning
Corning® Polypropylene Bottles, Corning
Corning® Polypropylene Bottles, Corning
Wide Mouth, 500 ml, PP, 45
Price
$179.35
New Customer Promotional Price
Log in for availability
Case of 12 Each
Product Details & Documents
Bottles are manufactured from low density polyethylene and are offered with a narrow or wide mouth design.
  • Hollow glass stopper style
  • Reusable
  • Non sterile
The cap and dispensing tube are manufactured from polypropylene to optimize backflow of the solution. The design permits minimal turbulence of the liquid in the bottle.
Specifications
  • Capacité:
    500 ml
  • Désignation:
    Wide Mouth
  • Matière:
    PP
  • GL Thread Size:
    45